accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 897921-59-4

IUPAC Name: N-[4-(naphthalen-1-yl)phenyl]-[1,1'-biphenyl]-4-amine
Synonyms: N-[4-(naphthalen-1-yl)phenyl]-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 371.48
Molecular Formula: C28H21N
InChi Key: DOTSEUKJBPAPGG-UHFFFAOYSA-N
SMILES: N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results