Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[4-(1-Naphthyl)phenyl]-4-biphenylamine 98.0+%, TCI America™
SDP

Catalog No. N1053200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1053200MG 200 mg
N10531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1053200MG Supplier TCI America Supplier No. N1053200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 897921-59-4
Molecular Formula C28H21N
Molecular Weight (g/mol) 371.48
MDL Number MFCD28138088
InChI Key DOTSEUKJBPAPGG-UHFFFAOYSA-N
Synonym N-[4-(Naphthalen-1-yl)phenyl]-[1,1′-biphenyl]-4-amine
PubChem CID 58900802
IUPAC Name N-[4-(naphthalen-1-yl)phenyl]-[1,1'-biphenyl]-4-amine
SMILES N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 100°C
Quantity 200 mg
Formula Weight 371.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-[4-(1-Naphthyl)phenyl]-4-biphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.