accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89793-12-4

IUPAC Name: ethyl 2-chloropyrimidine-5-carboxylate
Synonyms: ethyl 2-chloropyrimidine-5-carboxylate
Molecular Weight (g/mol): 186.6
Molecular Formula: C7H7ClN2O2
InChi Key: IEMKQRSOAOPKRJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(Cl)N=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results