accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONO

CAS RN 898289-54-8

IUPAC Name: 6-(oxan-4-yloxy)pyridine-2-carbaldehyde
Synonyms: 6-(oxan-4-yloxy)pyridine-2-carbaldehyde
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: SBTGZJPHUYLRRU-UHFFFAOYSA-N
SMILES: O=CC1=NC(OC2CCOCC2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results