accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 898826-64-7

IUPAC Name: 2-[(6-{[4,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-3-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 2-[(6-{[4,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-3-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 610.56
Molecular Formula: C25H38O17
InChi Key: SADQXSYAYSUILX-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(CO)C(OC3OC(CO)C(OC4OC(CO)C(O)C(O)C4O)C(O)C3O)C(O)C2O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results