Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gb3-beta-MP 95.0+%, TCI America™
SDP

Catalog No. M1767100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1767100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1767100MG Supplier TCI America Supplier No. M1767100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 898826-64-7
Molecular Formula C25H38O17
Molecular Weight (g/mol) 610.562
InChI Key SADQXSYAYSUILX-VAAZCZNESA-N
Synonym 4-Methoxyphenyl O-(alpha-D-Galactopyranosyl)-(1-4)-O-(beta-D-galactopyranosyl)-(1-4)-beta-D-glucopyranoside
PubChem CID 90477358
IUPAC Name (2R,3R,4S,5R,6R)-2-[(2R,3R,4R,5R,6S)-6-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)OC3C(C(C(C(O3)CO)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 610.56
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gb3-beta-MP
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.