accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOBr

CAS RN 89901-03-1

IUPAC Name: methyl 4'-bromo-[1,1'-biphenyl]-4-carboxylate
Synonyms: methyl 4'-bromo-[1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol): 291.14
Molecular Formula: C14H11BrO2
InChi Key: JHNMJLKCONRGMK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results