accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CSSSSNCH3CH3

CAS RN 89964-93-2

IUPAC Name: N,N-diethyl{[(4-{[(diethylcarbamothioyl)sulfanyl]methyl}phenyl)methyl]sulfanyl}carbothioamide
Synonyms: N,N-diethyl{[(4-{[(diethylcarbamothioyl)sulfanyl]methyl}phenyl)methyl]sulfanyl}carbothioamide
Molecular Weight (g/mol): 400.68
Molecular Formula: C18H28N2S4
InChi Key: CKESHWQHGBKZBB-UHFFFAOYSA-N
SMILES: CCN(CC)C(=S)SCC1=CC=C(CSC(=S)N(CC)CC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results