Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylylenebis(N,N-diethyldithiocarbamate) 98.0+%, TCI America™
SDP

Catalog No. X00565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X00565G Supplier TCI America Supplier No. X00565G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89964-93-2
Molecular Formula C18H28N2S4
Molecular Weight (g/mol) 400.676
MDL Number MFCD00191696
InChI Key CKESHWQHGBKZBB-UHFFFAOYSA-N
Synonym 1,4-Bis(N,N-diethyldithiocarbamylmethyl)benzene, p-Xylenebis(N,N-diethyldithiocarbamate)
PubChem CID 590732
IUPAC Name [4-(diethylcarbamothioylsulfanylmethyl)phenyl]methyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SCC1=CC=C(C=C1)CSC(=S)N(CC)CC

Specifications

Melting Point 79°C
Color White
Quantity 5 g
Formula Weight 400.68
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material p-Xylylenebis(N,N-diethyldithiocarbamate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.