accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHO

CAS RN 90-01-7

IUPAC Name: 2-(hydroxymethyl)phenol
Synonyms: saligenin (ar)-salicyl alcohol (ar)-isomer salicyl alcohol salicyl alcohol saligenol Sal salicyl alcohol, (ar)-isomer 2-hydroxybenzylalkohol disodium salt salicyl alcohol monosodium salt salicyl alcohol
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: CQRYARSYNCAZFO-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1O
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (5)
Physical Form 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)