Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxybenzyl alcohol, 99%

Catalog No. AAA1441018
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1441018 50 g
AAA1441030 250 g
AAA144100B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1441018 Supplier Thermo Scientific Chemicals Supplier No. A1441018
  • $78.50
    $58.65 / Each of 1

    Save $19.85

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 90-01-7 | C7H8O2 | 124.14 g/mol

2-Hydroxybenzyl alcohol acts as a local anesthetic. It is used in gastrodin production via biotransformation by cultured cells of Aspergillus foetidus and Penicillium cyclopium and participates in the selective ring opening reaction of 4H-1,3,2-benzodioxasilines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzyl alcohol acts as a local anesthetic. It is used in gastrodin production via biotransformation by cultured cells of Aspergillus foetidus and Penicillium cyclopium and participates in the selective ring opening reaction of 4H-1,3,2-benzodioxasilines.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-01-7
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00004617
InChI Key CQRYARSYNCAZFO-UHFFFAOYSA-N
Synonym salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol
PubChem CID 5146
ChEBI CHEBI:16464
IUPAC Name 2-(hydroxymethyl)phenol
SMILES OCC1=CC=CC=C1O

Specifications

Melting Point 83°C to 87°C
Quantity 50 g
Beilstein 1907195
Merck Index 14,8325
Solubility Information Soluble in water.
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 2-Hydroxybenzyl alcohol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 201-960-5
RTECSNumber DO6430000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.