accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90-02-8

IUPAC Name: 2-hydroxybenzaldehyde
Synonyms: salicylaldehyde salicylaldehyd O-hydroxybenzaldehyde salicylaldehyde (product) salicylal salizylaldehyd salicyladehyde salicylaldehyde (substance)
Molecular Weight (g/mol): 122.12
Molecular Formula: C7H6O2
InChi Key: SMQUZDBALVYZAC-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C=O
Quantity 
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (7)
Boiling Point 
  • (5)
  • (3)
  • (2)
  • (5)
  • (4)
Physical Form 
  • (12)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (5)
  • (7)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (7)
  • (5)
  • (3)
  • (2)
  • (5)
  • (4)
  • (12)
  • (2)
  • (1)
  • (2)