Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Hydroxybenzaldehyde, MP Biomedicals

Catalog No. ICN206570 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206570 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206570 Supplier MP Biomedicals, Inc Supplier No. 0520657080

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90-02-8
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O

Specifications

Quantity 100 g
Formula Weight 122.12
Chemical Name or Material o-Hydroxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.