accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90-22-2

IUPAC Name: diethyl(methyl){2-[(3-methyl-2-phenylpentanoyl)oxy]ethyl}azanium bromide
Synonyms: valethamate bromide
Molecular Weight (g/mol): 386.37
Molecular Formula: C19H32BrNO2
InChi Key: CEJGGHKJHDHLAZ-UHFFFAOYNA-M
SMILES: [Br-].CCC(C)C(C(=O)OCC[N+](C)(CC)CC)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results