Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valethamate Bromide 98.0+%, TCI America™
SDP

Catalog No. V00631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V00631G Supplier TCI America Supplier No. V00631G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90-22-2
Molecular Formula C19H32BrNO2
Molecular Weight (g/mol) 386.37
MDL Number MFCD00031617
InChI Key CEJGGHKJHDHLAZ-UHFFFAOYNA-M
Synonym valethamate bromide, ediposin, epidosin, epidozin, resitan, murel, valethamate bromide jan, n,n-diethyl-n-methyl-2-3-methyl-2-phenylpentanoyl oxy ethanaminium bromide, diethyl methyl 2-3-methyl-2-phenylvaleryloxy ethyl ammonium bromide, 2-diethylaminoethyl 2-phenyl-3-methylvalerate methyl bromide
PubChem CID 7010
IUPAC Name diethyl(methyl){2-[(3-methyl-2-phenylpentanoyl)oxy]ethyl}azanium bromide
SMILES [Br-].CCC(C)C(C(=O)OCC[N+](C)(CC)CC)C1=CC=CC=C1

Specifications

Melting Point 127°C
Color White
Quantity 1 g
Formula Weight 386.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Valethamate Bromide
RTECSNumber BP7600000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.