accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90-39-1

N(S)(S)N(R)(S)

CAS RN 90-39-1

IUPAC Name: (1S,2R,9S,10S)-7,15-diazatetracyclo[7.7.1.0²,⁷.0¹⁰,¹⁵]heptadecane
Synonyms: sparteine l-sparteine 6β,7α,9α,11α-pachycarpine sparteine (inn) and its salts
Molecular Weight (g/mol): 234.39
Molecular Formula: C15H26N2
InChi Key: SLRCCWJSBJZJBV-ZQDZILKHSA-N
SMILES: C1CCN2C[C@@H]3C[C@@H](CN4CCCC[C@H]34)[C@@H]2C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)