CAS RN 90-89-1
IUPAC Name:
N,N-diethyl-4-methylpiperazine-1-carboxamide
Synonyms:
diethylcarbamazine camin carbamazine diethylcarbamazine citrate diethyl carbamazine hetrazan banocide notezine caracide caricide
Molecular Weight (g/mol):
199.3
Molecular Formula:
C10H21N3O
InChi Key:
RCKMWOKWVGPNJF-UHFFFAOYSA-N
SMILES:
CCN(CC)C(=O)N1CCN(C)CC1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Diethylcarbamazine
CAS: 90-89-1 Molecular Formula: C10H21N3O Molecular Weight (g/mol): 199.30 InChI Key: RCKMWOKWVGPNJF-UHFFFAOYSA-N IUPAC Name: N,N-diethyl-4-methylpiperazine-1-carboxamide SMILES: CCN(CC)C(=O)N1CCN(C)CC1