CAS RN 9000-01-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
5
of
5
results
1
2
Promotions Available
Acacia, Total ash <4%
Acacia, Total ash <4%
CAS: 9000-01-5 MDL Number: MFCD00081264 Synonym: Gum Arabic
3
4
Gum arabic, MP Biomedicals™
CAS: 9000-01-5 MDL Number: MFCD00081264
5
Acacia, Spray-Dried Powder, NF, Spectrum™ Chemical
CAS: 9000-01-5