accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COCH3OOOOOHCH3OOH3COCH3OOOOOHCH3OOH3COCH3OOOHOHOOHCH3OOH3COCH3OOOHOHOOH

CAS RN 9000-55-9

IUPAC Name: bis(16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one); bis(8-hydroxy-7-(hydroxymethyl)-5-(3,4,5-trimethoxyphenyl)-2H,5H,6H,7H,8H-naphtho[2,3-d][1,3]dioxole-6-carboxylic acid)
Synonyms: podophyllin
Molecular Weight (g/mol): 1693.67
Molecular Formula: C88H92O34
InChi Key: KOUZYZCESNTXJO-UHFFFAOYNA-N
SMILES: COC1=CC(=CC(OC)=C1OC)C1C2C(COC2=O)C(O)C2=CC3=C(OCO3)C=C12.COC1=CC(=CC(OC)=C1OC)C1C2C(COC2=O)C(O)C2=CC3=C(OCO3)C=C12.COC1=CC(=CC(OC)=C1OC)C1C(C(CO)C(O)C2=CC3=C(OCO3)C=C12)C(O)=O.COC1=CC(=CC(OC)=C1OC)C1C(C(CO)C(O)C2=CC3=C(OCO3)C=C12)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)