accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHNOOOHOHOH

CAS RN 9000-86-6

IUPAC Name: 1-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]-5-methyl-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 272.26
Molecular Formula: C11H16N2O6
InChi Key: VVJYUAYZJAKGRQ-UHFFFAOYNA-N
SMILES: CC1=CN(C2CC(O)C(O)C(CO)O2)C(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)