Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Glutamic pyruvic transaminase, porcine heart

Catalog No. AAJ62109JPL
Change view
Click to view available options
Quantity:
10 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62109JPL 10 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62109JPL Supplier Thermo Scientific Chemicals Supplier No. J62109JPL

It is used for research and clinical diagnostic assays for liver disease. Alanine transaminase (GPT/ALT) is used in multi-analyte clinical chemistry controls and calibrators for major IVD manufacturers. The alanine aminotransferase (ALT) blood test is typically used to detect liver injury. Some of the other applications include Diagnostic Controls, calibrators & standards, immunoassays, clinical chemistry, testing/assay validation, life Science, manufacturing, validation Studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-86-6
Molecular Formula C11H16N2O6
Molecular Weight (g/mol) 272.257
MDL Number MFCD00131192
InChI Key VVJYUAYZJAKGRQ-UHFFFAOYSA-N
Synonym gpt, 1-2-deoxyhexopyranosyl-5-methylpyrimidine-2,4 1h,3h-dione, 1-4,5-dihydroxy-6-hydroxymethyl oxan-2-yl-5-methylpyrimidine-2,4-dione, 1-4,5-dihydroxy-6-hydroxymethyl oxan-2-yl-5-methyl-3h-pyrimidine-2,4-dione, 1-4,5-dihydroxy-6-hydroxymethyl oxan-2-yl-5-methyl-pyrimidine-2,4-dione
PubChem CID 251717
IUPAC Name 1-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C(=O)NC1=O)C2CC(C(C(O2)CO)O)O

Specifications

Packaging Plastic Bottle
Beilstein 3562140
Solubility Information Dissolves readily at 5mg/ml in 0.05M potassium phosphate pH 7.5 to give a clear,pale-green solution. Soluble in distilled water or dilute buffer.
Quantity 10 kU
Physical Form Lyophilized Powder
Chemical Name or Material Glutamic pyruvic transaminase
EINECSNumber 232-561-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.