accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90000-25-2

IUPAC Name: 4-(3-aminopropyl)-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-(3-aminopropyl)-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 192.28
Molecular Formula: C7H16N2O2S
InChi Key: FSRMWCVZAUEPEQ-UHFFFAOYSA-N
SMILES: NCCCN1CCS(=O)(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results