CAS RN 9001-05-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MP Biomedicals™ Catalase from Bovine Liver
Natural antioxidant used to study roles of reactive oxygen species in gene expression and apoptosis
2