CAS RN 9001-73-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2
Thermo Scientific Chemicals Papain, Carica Papaya Latex
CAS: 9001-73-4 Synonym: EC 3.4.22.2