accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOOHOONHN(S)H3CH3CCH3CH3CH3

CAS RN 900152-72-9

IUPAC Name: 2-[(2S)-4-(tert-butoxy)-N-[(2,4-dimethoxyphenyl)methyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanamido]acetic acid
Synonyms: [(2S)-4-(tert-butoxy)-N-[(2,4-dimethoxyphenyl)methyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanamido]acetic acid
Molecular Weight (g/mol): 618.68
Molecular Formula: C34H38N2O9
InChi Key: SGFXBTBQHJNOAB-NDEPHWFRSA-N
SMILES: COC1=CC(OC)=C(CN(CC(O)=O)C(=O)[C@H](CC(=O)OC(C)(C)C)NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)