accessibility menu, dialog, popup

UserName

Viewing profile as user:
∗∗ONH2n

CAS RN 9003-05-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C3H5NO)n
InChi Key: —
SMILES: NC(=O)C(-*)C-*
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)