Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(acrylamide) Solution Average Mw ~10 000 50 Wt. % In H2o

Catalog No. 91269725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-269-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-269-725G Supplier Sigma Aldrich Supplier No. 91269725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(acrylamide) is a water-soluble polymer made up of acrylamide subunits. Polyacrylamides have been reported to be protein resistant and biocompatible. Polyacrylamides have been used as carriers for delivery of drugs and bioactive compounds, or to as tissue engineering materials. Additionally, polycacrylamides have been used to increase the viscosity of water and facilitate the flocculation of particles present in water.

Specifications

CAS 9003-05-8
Linear Formula (C3H5NO)n
Quantity 25 g
Synonym PAM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.