accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90030-48-1

IUPAC Name: sodium 3-methoxy-3-oxopropane-1-sulfinate
Synonyms: sodium 3-methoxy-3-oxopropane-1-sulfinate
Molecular Weight (g/mol): 174.15
Molecular Formula: C4H7NaO4S
InChi Key: YICFALDKKMHFLT-UHFFFAOYSA-M
SMILES: [Na+].COC(=O)CCS([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results