Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Methyl 3-Sulfinopropionate 96.0+%, TCI America™
SDP

Catalog No. S0934200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0934200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0934200MG Supplier TCI America Supplier No. S0934200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90030-48-1
Molecular Formula C4H7NaO4S
Molecular Weight (g/mol) 174.146
MDL Number MFCD07776796
InChI Key YICFALDKKMHFLT-UHFFFAOYSA-M
Synonym 3-Sulfinopropionic Acid 1-Methyl Ester Sodium Salt, Sodium 3-Methoxy-3-oxopropane-1-sulfinate, 3-Methoxy-3-oxopropane-1-sulfinic Acid Sodium Salt
PubChem CID 23665669
IUPAC Name sodium;3-methoxy-3-oxopropane-1-sulfinate
SMILES COC(=O)CCS(=O)[O-].[Na+]

Specifications

Melting Point 207°C
Quantity 200 mg
Formula Weight 174.15
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Methyl 3-Sulfinopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.