accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNHNCH3

CAS RN 90038-01-0

IUPAC Name: hydrogen 5-[(2,3-dimethylphenyl)methyl]-1H-imidazole chloride
Synonyms: hydrogen detomidine chloride
Molecular Weight (g/mol): 222.72
Molecular Formula: C12H15ClN2
InChi Key: OIWRDXKNDCJZSM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=CC(CC2=CN=CN2)=C1C
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results