Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Detomidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D4331200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4331200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4331200MG Supplier TCI America Supplier No. D4331200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent with ≥98.0%(T)(HPLC)

Chemical Identifiers

CAS 90038-01-0
Molecular Formula C12H15ClN2
Molecular Weight (g/mol) 222.716
MDL Number MFCD17292784
InChI Key OIWRDXKNDCJZSM-UHFFFAOYSA-N
Synonym 5-(2,3-Dimethylbenzyl)-1H-imidazole Hydrochloride
PubChem CID 56031
IUPAC Name 5-[(2,3-dimethylphenyl)methyl]-1H-imidazole;hydrochloride
SMILES CC1=C(C(=CC=C1)CC2=CN=CN2)C.Cl

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 200 mg
Packaging Glass Bottle
Physical Form Crystal-Powder at 20°C
Formula Weight 222.72
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Chemical Name or Material Detomidine Hydrochloride
RTECSNumber NI5160000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.