accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 9004-61-9

IUPAC Name: sodium (2S,3S,4R,5R,6R)-3-{[(2S,3R,4R,5S,6R)-4-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3-acetamido-5-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-6-{[(2R,3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4,5-dihydroxyoxane-2-carboxylic acid
Synonyms: sodium hyaluronate
Molecular Weight (g/mol): 799.64
Molecular Formula: C28H44N2NaO23
InChi Key: YWIVKILSMZOHHF-QJZPQSOGSA-N
SMILES: [Na+].CC(=O)N[C@H]1[C@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O[C@@H]1O[C@@H]([C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)[C@H]2NC(C)=O)[C@H](O)[C@H]1O)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)