Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyaluronic acid, bovine vitreous humor

Catalog No. AAJ60566MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60566MF 50 mg
AAJ60566MA 10 mg
AAJ60566MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60566MF Supplier Thermo Scientific Chemicals Supplier No. J60566MF
  • $826.00
    $652.65 / Each of 1

    Save $173.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

Hyaluronic acid from bovine vitreous humor, CAS # 9004-61-9, is a naturally derived glycosaminoglycan that shows an angiogenic activity that promotes wound healing processes. | CAS: 9004-61-9 | C28H44N2O23 | 776.651 g/mol

Hyaluronic acid is a naturally derived, non - immunogenic, non - adhesive glycosaminoglycan that plays a prominent role in various wound - healing processes, as it as it is naturally angiogenic when degraded to small fragments. Hyaluronic acid promotes early inflammation which is critical for initiating wound healing, but then moderates later stages of the process, allowing matrix stabilization and reduction of long term inflammation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hyaluronic acid, derived from bovine vitreous humor, is a naturally derived, non-immunogenic, non-adhesive glycosaminoglycan that shows an angiogenic activity promoting wound-healing processes when degraded to small fragments

Application

  • It is considered a mucopolysaccharide composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine, which forms the core of complex proteoglycan aggregates found in the extracellular matrix
  • It has been used to study the activation of the MAPK/ERK pathway following infection of macrophages with Leishmania amazonensis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-61-9
Molecular Formula C28H44N2O23
Molecular Weight (g/mol) 776.651
MDL Number MFCD00131348
InChI Key KIUKXJAPPMFGSW-YXBJCWEESA-N
Synonym hyaluronic acid, dsstox_cid_26750, dsstox_rid_81875, dsstox_gsid_46750, 4xi-alpha-d-xylo-hexopyranuronosyl-1->3-3xi-2-acetylamino-2-deoxy-beta-d-ribo-hexopyranosyl-1->4-5xi-beta-d-xylo-hexopyranuronosyl-1->3-2-acetylamino-2-deoxy-d-glucopyranose
PubChem CID 24728612
IUPAC Name (2S,4S,5R,6S)-6-[(2S,3R,5S,6R)-3-acetamido-2-[(3S,4R,5R,6R)-6-[(3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-2-carboxy-4,5-dihydroxyoxan-3-yl]oxy-5-hydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)OC2C(C(C(C(O2)C(=O)O)OC3C(C(C(C(O3)CO)O)OC4C(C(C(C(O4)C(=O)O)O)O)O)NC(=O)C)O)O

Specifications

Color White
Quantity 50 mg
Merck Index 14,4757
Solubility Information Soluble in water.
Physical Form Lyophilized Powder
Chemical Name or Material Hyaluronic acid, bovine vitreous humor
EINECSNumber 232-678-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.