accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)(S)(R)O(R)OO(S)OO(R)O∗H3COH(R)O(R)H3COHCH3OH(R)OOCH3OHO∗HOCH3CH3OHn

CAS RN 9004-64-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C24H44O16)n
InChi Key: —
SMILES: CC(O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)O)OC(-*)[C@H](OC(C)O)[C@H]2OC(C)O)[C@H](OC(C)O)[C@@H](OC(C)O)[C@@H]1O-*
Quantity 
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (5)
  • (3)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
Physical Form 
  • (5)
  • (8)
  • (4)
Grade 
  • (1)
  • (8)
Percent Purity 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (8)
  • (1)
  • (11)
  • (8)
  • (5)
  • (8)
  • (8)
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (5)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (5)
  • (8)
  • (4)
  • (1)
  • (8)
  • (8)