Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydroxypropyl cellulose

Catalog No. 4350075G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-500-75G 5 g
43-500-7250G 250 g
43-500-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 43-500-75G Supplier Sigma Aldrich Supplier No. 4350075G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydroxypropyl cellulose (HPC) is biocompatible in nature and exhibits low critical solution temperature (LCST). It has been reported to be prepared by reacting cellulose with alkali, producing alkali cellulose which is consecutively reacted with propylene oxide. HPC is soluble in water at lower than RT. Viscosity behaves inversely to rise in temperature.Copolymers composed of HPC backbones with poly((2-dimethyl amino)ethyl methacrylate side chains has been synthesized for gene delivery. HPC has been used as a binder in to nanostructured anatase membranes. Polymer conjugates made of HPC backbones and poly(N-isopropylacryamide) as side chains act as smart hydrogels. HPC has been reported to be studied as a glue.

Specifications

CAS 9004-64-2
Density 0.5 g/mL (at 25°C)
MDL Number MFCD00132688
Quantity 5 g
Molecular Weight (g/mol) 10000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.