accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 9005-00-9

IUPAC Name: [2-(octadecyloxy)ethyl]oxidanyl
Synonyms: [2-(octadecyloxy)ethyl]oxidanyl
Molecular Weight (g/mol): 313.55
Molecular Formula: C20H41O2
InChi Key: PSEGVHKUPXNGGJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOCC[O]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)