Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij™ 78, Thermo Scientific™

Catalog No. AC216745000
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC216745000 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC216745000 Supplier Thermo Scientific Chemicals Supplier No. 216745000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9005-00-9
Acid Value 1mg KOH/g max.
Molecular Formula C20H42O2
Linear Formula C18H37(OCH2CH2)20OH
MDL Number MFCD00074177
Quantity 500g
Merck Index 10, 7449
Synonym Polyoxyethylene(20) stearyl ether
InChI Key ICIDSZQHPUZUHC-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCOCCO
IUPAC Name 2-octadecoxyethanol
Molecular Weight (g/mol) 314.554
Hydroxyl Value 45 to 60mg KOH/g
PubChem CID 75050
Formula Weight 1151.56
Infrared Spectrum Authentic
Water 3% max.
Chemical Name or Material Brij(r) 78
Show More Show Less
RTECSNumber WI6300000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.