accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 9007-73-2

IUPAC Name: hexadecane-1-sulfinate
Synonyms: hexadecane-1-sulfinate
Molecular Weight (g/mol): 289.5
Molecular Formula: C16H33O2S
InChi Key: HBTBNXFVJYRYGI-UHFFFAOYSA-M
SMILES: CCCCCCCCCCCCCCCCS([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)