Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferritin, Equine, MP Biomedicals™

Catalog No. MP215111990 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215111990 500 mg
MP215111980 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215111990 Supplier MP Biomedicals, Inc Supplier No. 0215111990
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Plays a key role in iron metabolism. Ferritin, Equine, MP Biomedicals is a ubiquitous iron storage protein with a shell of 24 subunits (apoferritin). It serves as an intracellular iron reserve (particularly in spleen, liver, intestinal mucosa, bone marrow

A shell of 24 protein subunits (apoferritin) and a core of Fe3+ ions with a peptide molecular weight of 440 kDa. The ferritin-like superfamily of iron-storage proteins is virtually ubiquitous in life on earth.

  • Studies have shown that ferritin iron incorporation is mediated by a ferroxidase activity associated with ferritin H subunits and a nucleation center associated with ferritin L subunits.
  • Release of iron from ferritin has an essential role in iron-dependent lipid peroxidation and may contribute to free radical-induced cell damage in vivo.
  • Therefore, by binding iron, ferritin may function as an antioxidant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9007-73-2
Molecular Formula C16H33O2S-
Molecular Weight (g/mol) 289.498
InChI Key HBTBNXFVJYRYGI-UHFFFAOYSA-M
Synonym 1exw, hsf
PubChem CID 6419224
IUPAC Name hexadecane-1-sulfinate
SMILES CCCCCCCCCCCCCCCCS(=O)[O-]

Specifications

Quantity 500 mg
Solubility Information Soluble in 0.15M sodium chloride, distilled water or dilute buffer.
Formula Weight ∼450,000 (Lit.)
Physical Form Viscous Liquid
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.