accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90090-64-5

IUPAC Name: (2R)-2-(thiophen-2-yl)pyrrolidin-1-ium
Synonyms: (2R)-2-(thiophen-2-yl)pyrrolidin-1-ium
Molecular Weight (g/mol): 154.25
Molecular Formula: C8H12NS
InChi Key: BNUVOMNEUJWHCF-SSDOTTSWSA-O
SMILES: C1C[NH2+][C@H](C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results