Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Thienyl)pyrrolidine, 97%, Thermo Scientific™

Catalog No. AAH5017903
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5017903 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5017903 Supplier Thermo Scientific Chemicals Supplier No. H5017903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(2-Thienyl)pyrrolidine is used in the preparation of imidazopyridine derivatives for treating viral infections.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 90090-64-5
Molecular Formula C8H11NS
Quantity 1g
MDL Number MFCD00052889
Sensitivity Moisture sensitive
Synonym 2-thiophen-2-yl pyrrolidine, 2-2-thienyl pyrrolidine, 2-thien-2-ylpyrrolidine, 2-thien-2-yl pyrrolidine, 2-thiophen-2-yl-pyrrolidine, pyrrolidine, 2-2-thienyl, 2-pyrrolidin-2-ylthiophene, akos bb-8869, acmc-209r4u, 3-thiophen-2-yl pyrrolidine
Solubility Information Soluble in water.
InChI Key BNUVOMNEUJWHCF-UHFFFAOYSA-N
SMILES C1CC(NC1)C2=CC=CS2
IUPAC Name 2-thiophen-2-ylpyrrolidine
Molecular Weight (g/mol) 153.243
PubChem CID 339554
Formula Weight 153.25
Percent Purity 97%
Chemical Name or Material 2-(2-Thienyl)pyrrolidine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.