accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNCl

CAS RN 90097-52-2

IUPAC Name: 2-chloro-3-(chloromethyl)quinoline
Synonyms: 2-chloro-3-(chloromethyl)quinoline
Molecular Weight (g/mol): 212.07
Molecular Formula: C10H7Cl2N
InChi Key: UOIJHUIOTFDGNA-UHFFFAOYSA-N
SMILES: ClCC1=C(Cl)N=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)