accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH2COOO

CAS RN 9011-16-9

IUPAC Name: 2,5-dihydrofuran-2,5-dione; methoxyethene
Synonyms: pvm/MA copolymer
Molecular Weight (g/mol): 156.14
Molecular Formula: C7H8O4
InChi Key: UPBDXRPQPOWRKR-UHFFFAOYSA-N
SMILES: COC=C.O=C1OC(=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results