CAS RN 9016-45-9
IUPAC Name:
2-[2-(4-nonylphenoxy)ethoxy]ethan-1-ol
Synonyms:
2-[2-(4-nonylphenoxy)ethoxy]ethanol
Molecular Weight (g/mol):
308.46
Molecular Formula:
C19H32O3
InChi Key:
BLXVTZPGEOGTGG-UHFFFAOYSA-N
SMILES:
CCCCCCCCCC1=CC=C(OCCOCCO)C=C1
1
–
2
of
2
results
2
MilliporeSigma™ NP-40 Alternative, Calbiochem™,
CAS: 9016-45-9 Molecular Formula: C19H32O3 Molecular Weight (g/mol): 308.46 MDL Number: MFCD00134080 InChI Key: BLXVTZPGEOGTGG-UHFFFAOYSA-N Synonym: Nonylphenyl Polyethylene Glycol PubChem CID: 24773 IUPAC Name: 2-[2-(4-nonylphenoxy)ethoxy]ethan-1-ol SMILES: CCCCCCCCCC1=CC=C(OCCOCCO)C=C1