Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NP-40 Alternative, Calbiochem™,

Catalog No. 492016500ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
49-201-6500ML 500 mL Glass Bottle
49-201-6100ML 100 mL Glass Bottle
49-201-61000M 1000 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 49-201-6500ML Supplier MilliporeSigma™ Supplier No. 492016500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9016-45-9
Molecular Formula C19H32O3
Molecular Weight (g/mol) 308.46
MDL Number MFCD00134080
InChI Key BLXVTZPGEOGTGG-UHFFFAOYSA-N
Synonym Nonylphenyl Polyethylene Glycol
PubChem CID 24773
IUPAC Name 2-[2-(4-nonylphenoxy)ethoxy]ethan-1-ol
SMILES CCCCCCCCCC1=CC=C(OCCOCCO)C=C1

Specifications

Color Undesignated
Quantity 500 mL
Packaging Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.