accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3MgBr

CAS RN 90224-21-8

IUPAC Name: bromo(2-ethylhexyl)magnesium
Synonyms: bromo(2-ethylhexyl)magnesium
Molecular Weight (g/mol): 217.43
Molecular Formula: C8H17BrMg
InChi Key: NDCJRLBSQQDNOO-UHFFFAOYNA-M
SMILES: CCCCC(CC)C[Mg]Br
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)