Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di(1-Adamantyl)Benzylphosphine

Catalog No. 6718001G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-180-01G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-180-01G Supplier Sigma Aldrich Supplier No. 6718001G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

sold in collaboration with Solvias AGCatalyst for:Sonogashira coupling reactionRuthenium-catalyzed amination of secondary alcohols. with ammoniaRhodium-catalyzed regioselective hydroformylation of alkenesReductive carbonylationSuzuki coupling reactioncataCXium is a registered trademark of Evonik Degussa GmbH

Specifications

CAS 90224-21-8
Density 0.863 g/mL at 25°C
Molecular Formula C8H17BrMg
Linear Formula C8H17BrMg
Quantity 100 mL
MDL Number MFCD02260220
Molecular Weight (g/mol) 217.43 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.