accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 902454-26-6

IUPAC Name: 4-[(1-methylpiperidin-4-yl)methoxy]aniline
Synonyms: 4-[(1-methylpiperidin-4-yl)methoxy]aniline
Molecular Weight (g/mol): 220.32
Molecular Formula: C13H20N2O
InChi Key: WZRKMUXDLMLRJV-UHFFFAOYSA-N
SMILES: CN1CCC(COC2=CC=C(N)C=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results