CAS RN 9028-00-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Clostripain, Clostridium histolyticum, Thermo Scientific Chemicals
CAS: 9028-00-6 MDL Number: MFCD00130814 Synonym: Endoproteinase-Arg-C; Clostridiopeptidase b