Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostripain, Clostridium histolyticum, Thermo Scientific Chemicals

Catalog No. AAJ61362MCR
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61362MCR 1 mg
AAJ61362MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61362MCR Supplier Thermo Scientific Chemicals Supplier No. J61362MCR
Add to Cart

A protease that cleaves proteins on the carboxyl bond of arginine

A protease that cleaves proteins on the carboxyl bond of arginine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9028-00-6
MDL Number MFCD00130814
Synonym Endoproteinase-Arg-C; Clostridiopeptidase b

Specifications

Color White
Quantity 1 mg
Physical Form Powder
Chemical Name or Material Clostripain, Clostridium histolyticum
How many units of clostripain are in one milligram (dry weight) of Clostripain, Clostridium histolyticum?

Each mg of Clostripain, Clostridium histolyticum (dry weight) contains a minimum of 50.0 units of clostripain.

Does Clostripain, Clostridium histolyticum, contain a His Tag?

The clostripain in Clostripain, Clostridium histolyticum is the native protein and does not have a His Tag.

Hazard Category H315-H319-H334-H335
Precautionary Statement P261-P264b-P271-P280-P285-P302+P352-P304+P340-P305+P351+P338-P332+P313-P342+P311-P362-P501c
EINECSNumber 232-822-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.